Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APG12B
Entrez Gene ID
820611
Gene Name
Ubiquitin-like superfamily protein
Gene Aliases
APG12, AT3G13970, ATG12B, AUTOPHAGY 12, AUTOPHAGY 12 B, AUTOPHAGY 12B
Chromosome Location
Chr.3: 4613492 - 4614857 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338096.1 NP_001326255.1 3 - 4 210 86
NM_112251.4 NP_188013.2 3 - 4 219 86
GenBank mRNA AB073185.1 - 3 - 4 163 86
AK118997.1 - 3 - 4 210 86
BT024653.1 - 3 - 4 156 86

View Full Product Details