Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSL5
Entrez Gene ID
820710
Gene Name
mechanosensitive channel of small conductance-like 5
Gene Aliases
AT3G14810, mechanosensitive channel of small conductance-like 5
Chromosome Location
Chr.3: 4970944 - 4974534 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202966.1 NP_001189895.1 4 - 5 2354 123
NM_112342.4 NP_188099.2 3 - 4 2446 123

View Full Product Details