The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036700.2 | NP_001031777.1 | 1 - 2 | 158 | 103 |
NM_119425.5 | NP_567904.1 | 1 - 2 | 173 | 103 | |
GenBank mRNA | AY056237.1 | - | 1 - 2 | 97 | 103 |