Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCA5
Entrez Gene ID
829498
Gene Name
beta carbonic anhydrase 5
Gene Aliases
A. THALIANA BETA CARBONIC ANHYDRASE 5, AT4G33580, ATBCA5, T16L1.70, T16L1_70, beta carbonic anhydrase 5
Chromosome Location
Chr.4: 16138251 - 16141624 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036707.1 NP_001031784.1 6 - 7 657 94
NM_001342222.1 NP_001328574.1 5 - 6 671 94
NM_119514.4 NP_567928.1 6 - 7 763 94
GenBank mRNA AK221870.1 - 6 - 7 655 94
AY034930.1 - 5 - 6 518 94
AY088420.1 - 5 - 6 555 94
AY122986.1 - 5 - 6 463 94

View Full Product Details