Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DML3
Entrez Gene ID
829552
Gene Name
demeter-like protein 3
Gene Aliases
AT4G34060, F28A23.180, F28A23_180, demeter-like protein 3
Chromosome Location
Chr.4: 16313366 - 16319426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342250.1 NP_001329149.1 19 - 20 3086 126
NM_001342251.1 NP_001329150.1 16 - 17 3111 126
NM_001342252.1 NP_001329148.1 17 - 18 2959 126
NM_119567.4 NP_195132.3 18 - 19 3013 126

View Full Product Details