Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SULTR1;2
Entrez Gene ID
844135
Gene Name
sulfate transporter 1;2
Gene Aliases
AT1G78000, F28K19.22, F28K19_22, SEL1, SELENATE RESISTANT 1, SULFATE TRANSPORTER, sulfate transporter 1;2
Chromosome Location
Chr.1: 29327515 - 29333220 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334809.1 NP_001321366.1 10 - 11 2541 89
NM_001334810.1 NP_001321365.1 10 - 11 1953 89
NM_001334811.1 NP_001321364.1 10 - 11 2127 89
NM_001334813.1 NP_001321362.1 8 - 9 1407 89
NM_106449.3 NP_565166.1 10 - 11 2127 89
NM_179568.2 NP_849899.1 9 - 10 1959 89
GenBank mRNA AB042322.2 - 10 - 11 1721 89
AK227518.1 - 9 - 10 1706 89

View Full Product Details