Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CALS5
- Entrez Gene ID
-
815852
- Gene Name
- callose synthase 5
- Gene Aliases
-
ARABIDOPSIS THALIANA GLUCAN SYNTHASE-LIKE 2, AT2G13680, ATGSL02, GLS2, GLUCAN SYNTHASE-LIKE 2, callose synthase 5
- Chromosome Location
-
Chr.2: 5694381 - 5706442 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_179622.4 |
NP_849953.2 |
7 - 8 |
792 |
63 |
View Full Product Details