Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPT1
Entrez Gene ID
816412
Gene Name
homogentisate phytyltransferase 1
Gene Aliases
AT2G18950, ATHPT, F19F24.15, F19F24_15, HOMOGENTISATE PHYTYLTRANSFERASE, TOCOPHEROL PHYTYLTRANSFERASE, TOCOPHEROL POLYPRENYLTRANSFERASE, TPT1, VITAMIN E 2, VTE2, homogentisate phytyltransferase 1
Chromosome Location
Chr.2: 8207199 - 8210513 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335615.1 NP_001325303.1 1 - 2 208 77
NM_179653.4 NP_849984.1 1 - 2 329 77
GenBank mRNA AF324344.1 - 1 - 2 151 77
AY063893.1 - 1 - 2 181 77
AY089963.1 - 1 - 2 37 77
AY113993.1 - 1 - 2 37 77
GU198365.1 - 1 - 2 115 77

View Full Product Details