Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPT1
Entrez Gene ID
816412
Gene Name
homogentisate phytyltransferase 1
Gene Aliases
AT2G18950, ATHPT, F19F24.15, F19F24_15, HOMOGENTISATE PHYTYLTRANSFERASE, TOCOPHEROL PHYTYLTRANSFERASE, TOCOPHEROL POLYPRENYLTRANSFERASE, TPT1, VITAMIN E 2, VTE2, homogentisate phytyltransferase 1
Chromosome Location
Chr.2: 8207199 - 8210513 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335615.1 NP_001325303.1 9 - 10 1030 109
NM_179653.4 NP_849984.1 9 - 10 1151 109
GenBank mRNA AF324344.1 - 9 - 10 973 109
AY063893.1 - 9 - 10 1003 109
AY089963.1 - 9 - 10 859 109
AY113993.1 - 9 - 10 859 109
GU198365.1 - 9 - 10 937 109

View Full Product Details