Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCL2
Entrez Gene ID
816606
Gene Name
GCR2-like 2
Gene Aliases
AT2G20770, F5H14.26, F5H14_26, GCR2-like 2
Chromosome Location
Chr.2: 8945243 - 8947310 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179672.4 NP_850003.1 4 - 5 699 103
GenBank mRNA AY063934.1 - 4 - 5 624 103
AY096480.1 - 4 - 5 562 103

View Full Product Details