Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCRK
Entrez Gene ID
817371
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT2G28250, T3B23.8, T3B23_8
Chromosome Location
Chr.2: 12042348 - 12046616 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036358.2 NP_001031435.1 2 - 3 718 85
NM_001336153.1 NP_001323735.1 2 - 3 451 85
NM_001336154.1 NP_001323736.1 2 - 3 192 85
NM_001336155.1 NP_001323737.1 2 - 3 577 85
NM_001336156.1 NP_001323738.1 2 - 3 500 85
NM_179784.2 NP_850115.1 1 - 2 687 85
GenBank mRNA AY065457.1 - 1 - 2 239 85
BT004417.1 - 1 - 2 69 85

View Full Product Details