Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYD
Entrez Gene ID
817375
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT2G28290, CHR3, CHROMATIN REMODELING COMPLEX SUBUNIT R, CHROMATIN REMODELING COMPLEX SUBUNIT R 3, SPLAYED
Chromosome Location
Chr.2: 12056213 - 12073279 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084502.2 NP_001077971.1 12 - 13 1902 94
NM_001336158.1 NP_001325347.1 11 - 12 1788 94
NM_001336159.1 NP_001325348.1 11 - 12 1791 94
NM_001336160.1 NP_001325349.1 11 - 12 1791 94
NM_179785.3 NP_850116.1 12 - 13 1902 94
NM_179786.2 NP_850117.1 12 - 13 1902 94

View Full Product Details