Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPT2
Entrez Gene ID
817601
Gene Name
Phototropic-responsive NPH3 family protein
Gene Aliases
AT2G30510, AT2G30520, ROOT PHOTOTROPISM 2, T6B20.13, T6B20_13
Chromosome Location
Chr.2: 13002686 - 13005746 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036369.2 NP_001031446.1 2 - 3 740 67
NM_001084513.2 NP_001077982.1 2 - 3 685 67
NM_179816.3 NP_850147.1 3 - 4 693 67
GenBank mRNA AK175297.1 - 2 - 3 735 67
AK221310.1 - 3 - 4 547 67
AK221993.1 - 3 - 4 547 67
AK222218.1 - 3 - 4 543 67
AK229759.1 - 3 - 4 542 67
AK229844.1 - 3 - 4 547 67
AK230435.1 - 3 - 4 542 67
AY035063.1 - 3 - 4 538 67
AY056359.1 - 3 - 4 520 67
BT000675.1 - 3 - 4 542 67

View Full Product Details