Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NAI2
Entrez Gene ID
820839
Gene Name
DNA topoisomerase-like protein
Gene Aliases
AT3G15950
Chromosome Location
Chr.3: 5397541 - 5402653 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035631.2 NP_001030708.1 14 - 15 942 85
NM_001338191.1 NP_001326809.1 14 - 15 942 85
NM_112465.4 NP_188216.2 16 - 17 1056 85
GenBank mRNA AY120750.1 - 16 - 17 1055 85
BT001207.1 - 16 - 17 1014 85

View Full Product Details