Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFP1
Entrez Gene ID
820845
Gene Name
MAR binding filament-like protein 1
Gene Aliases
AT3G16000, MAR binding filament-like protein 1
Chromosome Location
Chr.3: 5430758 - 5433851 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112470.3 NP_188221.2 1 - 2 525 79
GenBank mRNA BT008690.1 - 1 - 2 493 79

View Full Product Details