The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112555.2 | NP_566559.1 | 2 - 3 | 388 | 99 |
GenBank mRNA | AY045638.1 | - | 2 - 3 | 345 | 99 |