Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPSN13
Entrez Gene ID
821008
Gene Name
Putative plant snare 13
Gene Aliases
AT3G17440, ATNPSN13, novel plant snare 13
Chromosome Location
Chr.3: 5969564 - 5972572 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035639.2 NP_001030716.1 6 - 7 698 106
NM_112623.5 NP_566578.1 6 - 7 768 106
GenBank mRNA AY099622.1 - 6 - 7 636 106
BT001249.1 - 6 - 7 486 106

View Full Product Details