Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HVA22K
Entrez Gene ID
829825
Gene Name
HVA22-like protein K
Gene Aliases
AP22.45, AP22_45, AT4G36720, HVA22-like protein K
Chromosome Location
Chr.4: 17307709 - 17309867 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119836.3 NP_195390.2 2 - 3 181 90
GenBank mRNA BT012190.1 - 2 - 3 121 90

View Full Product Details