Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMP2
Entrez Gene ID
829866
Gene Name
Pre-mRNA splicing Prp18-interacting factor
Gene Aliases
AP22.39, AP22_39, AT4G37120, SWELLMAP 2
Chromosome Location
Chr.4: 17486516 - 17489354 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119875.4 NP_568017.1 2 - 3 340 128
GenBank mRNA AY056418.1 - 2 - 3 227 128

View Full Product Details