Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP60-2
Entrez Gene ID
817883
Gene Name
heat shock protein 60-2
Gene Aliases
AT2G33210, F25I18.5, F25I18_5, heat shock protein 60-2
Chromosome Location
Chr.2: 14074767 - 14078772 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202736.1 NP_001189665.1 9 - 10 1151 121
NM_179872.3 NP_850203.1 9 - 10 1194 121
GenBank mRNA AK175415.1 - 9 - 10 1153 121
AY136360.1 - 9 - 10 1030 121
BT000103.1 - 9 - 10 990 121

View Full Product Details