Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT2G34150
Entrez Gene ID
817976
Gene Name
SCAR family protein
Gene Aliases
AT2G34150, ATRANGAP2, ATSCAR1, SCAR1, T14G11.27, T14G11_27, WAVE1, WISKOTT-ALDRICH SYNDROME PROTEIN FAMILY VERPROLIN HOMOLOGOUS PROTEIN 1
Chromosome Location
Chr.2: 14419263 - 14423478 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036397.3 NP_001031474.1 5 - 6 821 68
NM_001336486.1 NP_001324986.1 2 - 3 455 68
NM_001336487.1 NP_001324987.1 4 - 5 498 68
NM_001336488.1 NP_001324988.1 3 - 4 494 68
NM_179894.2 NP_850225.1 5 - 6 847 68

View Full Product Details