Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE22
Entrez Gene ID
818043
Gene Name
maternal effect embryo arrest 22
Gene Aliases
AT2G34780, EMB1611, EMBRYO DEFECTIVE 1611, F19I3.1, F19I3_1, MATERNAL EFFECT EMBRYO ARREST 22
Chromosome Location
Chr.2: 14668440 - 14673904 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202750.1 NP_001189679.1 7 - 8 3920 65
NM_001336527.1 NP_001324225.1 6 - 7 3476 65
NM_001336528.1 NP_001324226.1 5 - 6 3428 65
NM_179906.3 NP_850237.1 7 - 8 3771 65

View Full Product Details