Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RA-5
Entrez Gene ID
837023
Gene Name
RAS 5
Gene Aliases
ARA-5, ARA5, ARABIDOPSIS RAS 5, ARABIDOPSIS THALIANA RAB D2A, ARABIDOPSIS THALIANA RESPONSIVE TO ABSCISIC ACID 1B, AT1G02130, ATRAB1B, ATRABD2A, RAB D2A, RABD2A, RAS 5, T7I23.6, T7I23_6
Chromosome Location
Chr.1: 399983 - 401919 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100093.4 NP_171715.1 5 - 6 388 138
GenBank mRNA AY052333.1 - 5 - 6 312 138
AY061905.1 - 4 - 5 257 138

View Full Product Details