Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPI
Entrez Gene ID
839323
Gene Name
BEACH-DOMAIN HOMOLOG A1
Gene Aliases
AT1G03060, F10O3.12, F10O3_12, SPIRRIG
Chromosome Location
Chr.1: 712440 - 727553 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331381.1 NP_001322129.1 10 - 11 2877 83
NM_001331382.1 NP_001322128.1 10 - 11 2890 83
NM_001331383.1 NP_001322130.1 10 - 11 2497 83
NM_100188.1 NP_171805.1 10 - 11 2231 83

View Full Product Details