Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRP4
Entrez Gene ID
839521
Gene Name
ribosomal RNA processing 4
Gene Aliases
AT1G03360, ATRRP4, F15K9.4, F15K9_4, ribosomal RNA processing 4
Chromosome Location
Chr.1: 824517 - 826399 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100218.5 NP_171835.1 4 - 5 533 81
GenBank mRNA AK176232.1 - 4 - 5 513 81
BT010533.1 - 4 - 5 397 81

View Full Product Details