Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SWI2
- Entrez Gene ID
-
839417
- Gene Name
- switch 2
- Gene Aliases
-
AT1G03750, CHR9, CHROMATIN REMODELING 9, F21M11.34, F21M11_34, SNF2, SUCROSE NON-FERMENTING 2, switch 2
- Chromosome Location
-
Chr.1: 937826 - 941162 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100254.4 |
NP_171871.2 |
3 - 4 |
1753 |
138 |
View Full Product Details