Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP88A3
Entrez Gene ID
839311
Gene Name
cytochrome P450, family 88, subfamily A, polypeptide 3
Gene Aliases
''cytochrome P450, AT1G05160, ATKAO1, ENT-KAURENOIC ACID OXYDASE 1, KAO1, YUP8H12.23, YUP8H12_23, cytochrome P450, family 88, polypeptide 3, polypeptide 3'', subfamily A
Chromosome Location
Chr.1: 1487378 - 1491005 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331521.1 NP_001320198.1 4 - 5 1309 97
NM_100394.4 NP_172008.1 4 - 5 1376 97
GenBank mRNA AK229009.1 - 4 - 5 1311 97
BT029169.1 - 3 - 4 975 97

View Full Product Details