Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP88A3
Entrez Gene ID
839311
Gene Name
cytochrome P450, family 88, subfamily A, polypeptide 3
Gene Aliases
''cytochrome P450, AT1G05160, ATKAO1, ENT-KAURENOIC ACID OXYDASE 1, KAO1, YUP8H12.23, YUP8H12_23, cytochrome P450, family 88, polypeptide 3, polypeptide 3'', subfamily A
Chromosome Location
Chr.1: 1487378 - 1491005 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331521.1 NP_001320198.1 7 - 8 1586 80
NM_100394.4 NP_172008.1 7 - 8 1653 80
GenBank mRNA AK229009.1 - 7 - 8 1588 80
BT029169.1 - 6 - 7 1252 80

View Full Product Details