Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARI12
Entrez Gene ID
837098
Gene Name
RING/U-box superfamily protein
Gene Aliases
ARABIDOPSIS ARIADNE 12, ARIADNE 12, AT1G05880, ATARI12, T20M3.15, T20M3_15
Chromosome Location
Chr.1: 1775643 - 1778553 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197990.1 NP_001184919.1 9 - 10 1060 110
NM_001331600.1 NP_001322817.1 9 - 10 1060 110
NM_100469.2 NP_172079.2 9 - 10 1039 110

View Full Product Details