Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP1A
Entrez Gene ID
819132
Gene Name
methionine aminopeptidase 1A
Gene Aliases
AT2G45240, F4L23.25, methionine aminopeptidase 1A
Chromosome Location
Chr.2: 18655760 - 18659111 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337118.1 NP_001325142.1 7 - 8 737 108
NM_130087.4 NP_182049.1 7 - 8 863 108
GenBank mRNA AF250960.1 - 7 - 8 619 108
AK227222.1 - 7 - 8 659 108
AY099542.1 - 7 - 8 634 108
BT008490.1 - 7 - 8 564 108

View Full Product Details