Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY46
Entrez Gene ID
819248
Gene Name
WRKY DNA-binding protein 46
Gene Aliases
AT2G46400, ATWRKY46, F11C10.9, WRKY DNA-BINDING PROTEIN 46, WRKY DNA-binding protein 46
Chromosome Location
Chr.2: 19043411 - 19044955 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130204.3 NP_182163.1 2 - 3 572 84
GenBank mRNA AY046275.1 - 2 - 3 371 84
AY080812.1 - 2 - 3 443 84
AY117210.1 - 2 - 3 371 84

View Full Product Details