Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF11
Entrez Gene ID
819264
Gene Name
auxin response factor 11
Gene Aliases
AT2G46530, F11C10.34, auxin response factor 11
Chromosome Location
Chr.2: 19104632 - 19108486 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036471.1 NP_001031548.1 10 - 11 1552 66
NM_130218.5 NP_182176.2 11 - 12 1850 66
NM_201972.1 NP_973701.1 11 - 12 1794 66
GenBank mRNA AK229924.1 - 11 - 12 1661 66
AY669791.1 - 10 - 11 1552 66

View Full Product Details