Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RKF3
Entrez Gene ID
819413
Gene Name
receptor-like kinase in in flowers 3
Gene Aliases
AT2G48010, T9J23.16, receptor-like kinase in in flowers 3
Chromosome Location
Chr.2: 19641351 - 19643899 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130368.4 NP_182322.1 1 - 1 1948 122
GenBank mRNA AF024650.1 - 1 - 1 1852 122
AY037237.1 - 1 - 1 1924 122

View Full Product Details