Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNL
Entrez Gene ID
837306
Gene Name
RNAligase
Gene Aliases
ARABIDOPSIS THALIANA RNA LIGASE, AT1G07910, ATRNL, AtRLG1, RNAligase, T6D22.1, T6D22_1, TRNA LIGASE
Chromosome Location
Chr.1: 2445437 - 2454645 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331741.1 NP_001318950.1 16 - 17 2156 71
NM_100665.3 NP_172269.2 16 - 17 2034 71
GenBank mRNA AK230326.1 - 16 - 17 2001 71

View Full Product Details