Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAP8
Entrez Gene ID
837536
Gene Name
amino acid permease 8
Gene Aliases
AT1G10010, ATAAP8, T27I1.3, T27I1_3, amino acid permease 8
Chromosome Location
Chr.1: 3265886 - 3268823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331877.1 NP_001322343.1 2 - 3 506 79
NM_100875.1 NP_172472.1 2 - 3 416 79

View Full Product Details