Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GDU4
Entrez Gene ID
817013
Gene Name
glutamine dumper 4
Gene Aliases
AT2G24762, AtGDU4, glutamine dumper 4
Chromosome Location
Chr.2: 10559173 - 10560239 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128035.5 NP_565577.1 1 - 1 906 113

View Full Product Details