Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RLP21
- Entrez Gene ID
-
817085
- Gene Name
- receptor like protein 21
- Gene Aliases
-
AT2G25470, AtRLP21, F13B15.13, F13B15_13, receptor like protein 21
- Chromosome Location
-
Chr.2: 10838294 - 10842016 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128104.5 |
NP_180117.4 |
6 - 7 |
970 |
69 |
View Full Product Details