Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ftsh4
Entrez Gene ID
817154
Gene Name
FTSH protease 4
Gene Aliases
AT2G26140, FTSH protease 4, T19L18.5, T19L18_5
Chromosome Location
Chr.2: 11131578 - 11135465 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128172.4 NP_565616.1 2 - 3 454 109

View Full Product Details