Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRV2
Entrez Gene ID
817230
Gene Name
DNAJ heat shock N-terminal domain-containing protein
Gene Aliases
AT2G26890, F12C20.7, F12C20_7, GRAVITROPISM DEFECTIVE 2, KAM2, KATAMARI2
Chromosome Location
Chr.2: 11462067 - 11474371 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128246.5 NP_180257.3 7 - 8 3871 102

View Full Product Details