Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AGC2-1
- Entrez Gene ID
-
822119
- Gene Name
- AGC (cAMP-dependent, cGMP-dependent and protein kinase C) kinase family protein
- Gene Aliases
-
AGC2, AGC2 kinase 1, AT3G25250, AtOXI1, OXI1, oxidative signal-inducible1
- Chromosome Location
-
Chr.3: 9195334 - 9197126 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113431.6 |
NP_189162.1 |
1 - 2 |
611 |
137 |
GenBank mRNA |
BT029732.1 |
- |
1 - 2 |
379 |
137 |
View Full Product Details