Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPH3
Entrez Gene ID
836554
Gene Name
Phototropic-responsive NPH3 family protein
Gene Aliases
AT5G64330, JK218, MSJ1.17, MSJ1_17, NON-PHOTOTROPIC HYPOCOTYL 3, ROOT PHOTOTROPISM 3, RPT3
Chromosome Location
Chr.5: 25727126 - 25730358 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345622.1 NP_001318873.1 1 - 2 436 85
NM_001345623.1 NP_001330808.1 1 - 2 436 85
NM_125829.4 NP_568989.1 1 - 2 578 85

View Full Product Details