Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEPKR1
Entrez Gene ID
837815
Gene Name
phosphoenolpyruvate carboxylase-related kinase 1
Gene Aliases
AT1G12580, F5O11.32, F5O11_32, phosphoenolpyruvate carboxylase-related kinase 1
Chromosome Location
Chr.1: 4282794 - 4285876 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101129.3 NP_172719.1 4 - 5 1638 78
GenBank mRNA AY057692.1 - 4 - 5 1533 78
AY143928.1 - 3 - 4 1284 78

View Full Product Details