Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VIK
Entrez Gene ID
837960
Gene Name
VH1-interacting kinase
Gene Aliases
AT1G14000, F7A19.9, F7A19_9, VH1-interacting kinase
Chromosome Location
Chr.1: 4797358 - 4800283 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101266.4 NP_172853.1 9 - 10 1320 73
GenBank mRNA BT026491.1 - 9 - 10 1072 73

View Full Product Details