Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ARC3
- Entrez Gene ID
-
843839
- Gene Name
- GTP binding protein
- Gene Aliases
-
ACCUMULATION AND REPLICATION OF CHLOROPLASTS 3, AT1G75010, F25A4.3, F25A4_3
- Chromosome Location
-
Chr.1: 28164828 - 28170113 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106158.5 |
NP_177638.2 |
6 - 7 |
912 |
143 |
View Full Product Details