Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CKX5
Entrez Gene ID
843881
Gene Name
cytokinin oxidase 5
Gene Aliases
ARABIDOPSIS THALIANA CYTOKININ OXIDASE 5, AT1G75450, ATCKX5, ATCKX6, CYTOKININ OXIDASE 6, F1B16.2, F1B16_2, cytokinin oxidase 5
Chromosome Location
Chr.1: 28314458 - 28318375 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198473.1 NP_001185402.1 3 - 4 1314 113
NM_106199.5 NP_177678.2 3 - 4 1298 113
GenBank mRNA AK176378.1 - 3 - 4 1298 113

View Full Product Details