Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC5A
Entrez Gene ID
844020
Gene Name
exocyst complex component sec5
Gene Aliases
AT1G76850, F7O12.2, F7O12_2, exocyst complex component sec5
Chromosome Location
Chr.1: 28847871 - 28854499 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106336.4 NP_177811.2 20 - 21 3197 114
GenBank mRNA AF479278.1 - 20 - 21 3117 114
AK118540.1 - 20 - 21 3108 114
BT005978.1 - 20 - 21 3055 114

View Full Product Details