Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC5A
Entrez Gene ID
844020
Gene Name
exocyst complex component sec5
Gene Aliases
AT1G76850, F7O12.2, F7O12_2, exocyst complex component sec5
Chromosome Location
Chr.1: 28847871 - 28854499 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106336.4 NP_177811.2 9 - 10 1612 104
GenBank mRNA AF479278.1 - 9 - 10 1532 104
AK118540.1 - 9 - 10 1523 104
BT005978.1 - 9 - 10 1470 104

View Full Product Details