Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5K2
Entrez Gene ID
844110
Gene Name
phosphatidylinositol-4-phosphate 5-kinase 2
Gene Aliases
AT1G77740, phosphatidylinositol-4-phosphate 5-kinase 2
Chromosome Location
Chr.1: 29220349 - 29224426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334792.1 NP_001319397.1 6 - 7 2145 101
NM_106423.2 NP_177897.1 6 - 7 1936 101

View Full Product Details