Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VHA-A
Entrez Gene ID
844228
Gene Name
vacuolar ATP synthase subunit A
Gene Aliases
AT1G78900, vacuolar ATP synthase subunit A
Chromosome Location
Chr.1: 29660088 - 29665078 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036222.3 NP_001031299.1 14 - 15 1567 126
NM_106539.6 NP_178011.1 15 - 16 1526 126
GenBank mRNA AK230248.1 - 15 - 16 1795 126
AK317358.1 - 14 - 15 1524 126
AY059909.1 - 15 - 16 1427 126
AY081296.1 - 15 - 16 1427 126
AY085759.1 - 15 - 16 1427 126
BT002589.1 - 14 - 15 1302 126
BT008383.1 - 14 - 15 1302 126
U65638.1 - 14 - 15 1330 126

Gene Symbol
AT5G21020
Entrez Gene ID
832226
Gene Name
transmembrane protein
Gene Aliases
AT5G21020
Chromosome Location
Chr.5: 7138762 - 7139661 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK230328.1 - - 934 126

View Full Product Details