Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ROPGEF12
- Entrez Gene ID
-
844325
- Gene Name
- RHO guanyl-nucleotide exchange factor 12
- Gene Aliases
-
AT1G79860, ATROPGEF12, F19K16.18, F19K16_18, KINASE PARTNER PROTEIN-LIKE, KPP-LIKE, MATERNAL EFFECT EMBRYO ARREST 64, MEE64, RHO guanyl-nucleotide exchange factor 12, ROP (rho of plants) guanine nucleotide exchange factor 12
- Chromosome Location
-
Chr.1: 30041989 - 30044297 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106635.4 |
NP_178104.1 |
3 - 4 |
821 |
73 |
GenBank mRNA |
BT033042.1 |
- |
3 - 4 |
616 |
73 |
View Full Product Details